Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam133b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042501.1 | NP_001035966.1 | 3 | 225 | ||
GenBank mRNA | AK008115.1 | - | 3 | 226 | ||
AK019188.1 | - | 3 | 226 | |||
AK020011.1 | - | 3 | 223 | |||
AK161505.1 | - | 3 | 166 | |||
BC027051.1 | - | 3 | 214 | |||
BC147310.1 | - | 3 | 220 | 9056359 | ||
BC147311.1 | - | 3 | 220 | 9056381 | ||
BC172033.1 | - | 3 | 220 | |||
BY711248.1 | - | 3 | 226 | |||
CK791263.1 | - | 2 | 43 |