Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026585.3 | NP_080861.2 | 3 | 270 | ||
GenBank mRNA | AK011156.1 | - | 3 | 216 | ||
AK038318.1 | - | 3 | 248 | |||
AK122320.1 | - | 2 | 276 | |||
AK145716.1 | - | 3 | 270 | |||
AK161181.1 | - | 3 | 219 | |||
AK163765.1 | - | 2 | 258 | |||
AK168388.1 | - | 3 | 257 | |||
AK215654.1 | - | 1 | 57 | |||
BC032177.1 | - | 1 | 497 | |||
BC040393.1 | - | 3 | 195 | |||
BC049979.1 | - | 3 | 246 | |||
BC056942.1 | - | 3 | 197 | 6827177 | ||
BY768668.1 | - | 3 | 271 |