Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000264.3 | NP_000255.2 | 6 | 1033 | ||
NM_001083602.1 | NP_001077071.1 | 6 | 1141 | |||
NM_001083603.1 | NP_001077072.1 | 6 | 987 | |||
NM_001083604.1 | NP_001077073.1 | 6 | 751 | |||
NM_001083605.1 | NP_001077074.1 | 6 | 734 | |||
NM_001083606.1 | NP_001077075.1 | 6 | 833 | |||
NM_001083607.1 | NP_001077076.1 | 6 | 907 | |||
GenBank mRNA | AB189436.1 | - | 6 | 1141 | ||
AB189437.1 | - | 6 | 987 | |||
AB189438.1 | - | 6 | 753 | |||
AB189439.1 | - | 6 | 736 | |||
AB189440.1 | - | 6 | 906 | |||
AB209495.1 | - | 3 | 482 | |||
DB093644.1 | - | 2 | 358 | |||
U43148.1 | - | 6 | 833 | |||
U59464.1 | - | 6 | 845 |