Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB186125.1 | - | 19, 20 | 4318 | ||
AK018850.1 | - | 2, 3 | 422 | |||
AK173252.1 | - | 16, 17 | 2646 | |||
BC031499.1 | - | 2, 3 | 258 | |||
BC039211.1 | - | 10, 11 | 1364 | |||
BC139006.1 | - | 17, 18 | 3790 | 8862028 | ||
BC158061.1 | - | 17, 18 | 3793 | 9007361 |