Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026485.2 | NP_080761.2 | 10 | 1905 | ||
GenBank mRNA | AB030206.1 | - | 1 | 317 | ||
AK014744.1 | - | 1 | 580 | |||
AK017745.1 | - | 10 | 1900 | |||
AK131116.1 | - | 2 | 3987 | |||
AK146027.1 | - | 10 | 1921 |