Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zwilch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026507.4 | NP_080783.3 | 9, 10 | 969 | ||
GenBank mRNA | AK009559.1 | - | 9, 10 | 940 | ||
AK012004.1 | - | 10, 11 | 1012 | |||
AK019825.1 | - | 9, 10 | 947 | |||
AK050917.1 | - | 9, 10 | 935 | |||
BC027435.1 | - | 9, 10 | 917 | 2812472 |