Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026521.4 | NP_080797.1 | 4 | 835 | ||
GenBank mRNA | AB041652.1 | - | 4 | 778 | ||
AK032231.1 | - | 4 | 721 | |||
AK082392.1 | - | 4 | 739 | |||
AK132053.1 | - | 4 | 752 | |||
AK145319.1 | - | 4 | 713 | |||
AK166530.1 | - | 4 | 713 | |||
AK167897.1 | - | 4 | 752 | |||
AK168958.1 | - | 4 | 714 | |||
AK215792.1 | - | 1 | 13 | |||
BC037600.1 | - | 4 | 760 | |||
BC042704.1 | - | 4 | 726 | |||
BC052459.1 | - | 4 | 835 | |||
BC054356.1 | - | 4 | 757 | |||
BC085477.1 | - | 4 | 719 | |||
BC147306.1 | - | 4 | 694 | 9056379 | ||
BC147307.1 | - | 4 | 694 | 9056367 |