Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142681.1 | NP_001136153.1 | 13 | 3329 | ||
NM_026522.5 | NP_080798.2 | 11 | 3071 | |||
GenBank mRNA | AK036717.1 | - | 13 | 3274 | ||
AK050655.1 | - | 11 | 3019 |