Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026538.3 | NP_080814.1 | 14 | 2683 | ||
GenBank mRNA | AK076975.1 | - | 14 | 2683 | ||
AK077953.1 | - | 9 | 1930 |