Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcakd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026551.3 | NP_080827.2 | 5 | 1699 | ||
GenBank mRNA | AK002835.1 | - | 2 | 1299 | ||
AK003426.1 | - | 1 | 1078 | |||
AK051022.1 | - | 5 | 1700 | |||
AK078460.1 | - | 5 | 1632 | |||
BC005607.1 | - | 5 | 1665 | 3498151 | ||
BC023208.1 | - | 4 | 1613 | |||
BC031855.1 | - | 5 | 1648 | |||
BC042685.1 | - | 4 | 1613 | |||
BC045193.1 | - | 5 | 1653 | 5368157 | ||
BC047340.1 | - | 4 | 1613 | |||
BC058591.1 | - | 1 | 1170 |