Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170485.1 | NP_001163956.1 | 3, 4 | 356 | ||
NM_001170486.1 | NP_001163957.1 | 4, 5 | 468 | |||
NM_026552.3 | NP_080828.1 | 5, 6 | 587 | |||
GenBank mRNA | AJ278129.1 | - | 5, 6 | 513 | ||
AK017993.1 | - | 5, 6 | 588 | |||
AK018069.1 | - | 5, 6 | 525 | |||
AK039215.1 | - | 5, 6 | 535 | |||
AK076248.1 | - | 5, 6 | 535 | |||
AK077325.1 | - | 5, 6 | 509 | |||
AK133626.1 | - | 5, 6 | 535 | |||
AK136082.1 | - | 5, 6 | 530 | |||
AK150690.1 | - | 5, 6 | 521 | |||
AK153380.1 | - | 5, 6 | 519 | |||
AK159410.1 | - | 4, 5 | 416 | |||
AK166440.1 | - | 5, 6 | 509 | |||
BC015280.1 | - | 4, 5 | 499 |