Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026570.4 | NP_080846.1 | 7 | 800 | ||
GenBank mRNA | AK002839.1 | - | 6 | 448 | ||
AK010522.1 | - | 7 | 801 | |||
AK012096.1 | - | 7 | 767 | |||
AK016290.1 | - | 2 | 236 | |||
AK081867.1 | - | 7 | 811 | |||
BC020043.1 | - | 7 | 768 | 4013548 |