Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000274.3 | NP_000265.1 | 2 | 278 | ||
NM_001322965.1 | NP_001309894.1 | 2 | 273 | |||
NM_001322966.1 | NP_001309895.1 | 3 | 593 | |||
NM_001322967.1 | NP_001309896.1 | 3 | 398 | |||
NM_001322968.1 | NP_001309897.1 | 4 | 485 | |||
NM_001322969.1 | NP_001309898.1 | 3 | 365 | |||
NM_001322970.1 | NP_001309899.1 | 4 | 581 | |||
NM_001322971.1 | NP_001309900.1 | 2 | 278 | |||
XM_017016279.1 | XP_016871768.1 | 2 | 220 | |||
GenBank mRNA | AB208817.1 | - | 2 | 209 | ||
AK312561.1 | - | 2 | 225 | |||
BC000964.2 | - | 2 | 206 | 3449883 | ||
BC016928.1 | - | 2 | 210 | 3913352 | ||
CD104356.1 | - | 2 | 221 | |||
CD683561.1 | - | 2 | 247 | |||
CR457045.1 | - | 1 | 145 | |||
DB459793.1 | - | 2 | 278 | |||
M12267.1 | - | 2 | 199 | |||
M14963.1 | - | 1 | 189 | |||
M23204.1 | - | 2 | 196 | |||
Y07511.1 | - | 1 | 185 |