Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000277.1 | NP_000268.1 | 2, 3 | 648 | ||
XM_017019370.1 | XP_016874859.1 | 2, 3 | 291 | |||
GenBank mRNA | AK298419.1 | - | 2, 3 | 272 | ||
AK313383.1 | - | 2, 4 | 273 | |||
BC026251.1 | - | 2, 3 | 329 | 4767736 | ||
K03020.1 | - | 2, 3 | 398 | |||
L47726.1 | - | 2, 3 | 648 | |||
U49897.1 | - | 2, 3 | 648 |