Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026627.1 | NP_080903.1 | 1 | 183 | ||
GenBank mRNA | AK006586.1 | - | 1 | 169 | ||
AK015538.1 | - | 1 | 174 | |||
AK018839.1 | - | 1 | 183 | |||
AK018883.1 | - | 1 | 105 | |||
AK018922.1 | - | 1 | 169 | |||
AK018951.1 | - | 1 | 169 | |||
AK018962.1 | - | 1 | 169 | |||
AK160415.1 | - | 1 | 169 | |||
BC048736.1 | - | 1 | 147 | |||
BC061062.1 | - | 1 | 195 | 6774311 | ||
EU079024.1 | - | 1 | 110 |