Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Toe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026654.2 | NP_080930.1 | 2 | 132 | ||
GenBank mRNA | AK016889.1 | - | 122 | |||
AK019829.1 | - | 2 | 132 | |||
AK046576.1 | - | 2 | 163 | |||
AK082975.1 | - | 2 | 113 | |||
BC023109.1 | - | 2 | 108 | 4021435 |