Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000287.3 | NP_000278.3 | 1 | 699 | ||
NM_001316313.1 | NP_001303242.1 | 1 | 699 | |||
NR_133009.1 | 1 | 699 | ||||
XM_011514661.1 | XP_011512963.1 | 1 | 699 | |||
GenBank mRNA | AB051076.1 | - | 1 | 606 | ||
AB051077.1 | - | 1 | 606 | |||
AB051078.1 | - | 1 | 606 | |||
AB208906.1 | - | 1 | 659 | |||
AK225704.1 | - | 1 | 637 | |||
AK314237.1 | - | 1 | 635 | |||
BC033487.1 | - | 1 | 614 | |||
BC048331.1 | - | 1 | 622 | 5189685 | ||
D83703.1 | - | 1 | 676 |