Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026701.2 | NP_080977.2 | 128 | |||
GenBank mRNA | AK002802.1 | - | 2 | 148 | ||
AK143947.1 | - | 2 | 164 | |||
BC019997.1 | - | 128 | 4240949 |