Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026784.3 | NP_081060.2 | 2 | 363 | ||
NM_027348.1 | NP_081624.1 | 2 | 332 | |||
GenBank mRNA | AK003607.1 | - | 2 | 166 | ||
AK013477.1 | - | 2 | 332 | |||
AK150483.1 | - | 2 | 282 | |||
AK168358.1 | - | 2 | 258 | |||
BY266316.1 | - | 2 | 365 |