Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaset2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083938.2 | NP_001077407.1 | 2, 3 | 267 | ||
GenBank mRNA | AK015947.1 | - | 2, 3 | 268 | ||
AK018722.1 | - | 2, 3 | 267 | |||
AK019234.1 | - | 2, 3 | 259 | |||
BC031496.1 | - | 2, 3 | 247 | 3156549 | ||
BC089534.1 | - | 2, 3 | 576 | 6703307 |
Gene Symbol: Rnaset2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026611.2 | NP_080887.1 | 2, 3 | 311 | ||
GenBank mRNA | AK029149.1 | - | 265 | |||
AK084454.1 | - | 271 | ||||
BC100330.1 | - | 2, 3 | 236 | 30527517 | ||
CK788185.1 | - | 2, 3 | 243 |