Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026615.4 | NP_080891.1 | 4 | 216 | ||
GenBank mRNA | AK012124.1 | - | 4 | 222 | ||
AK019352.1 | - | 4 | 222 | |||
AK049227.1 | - | 4 | 217 | |||
AK049849.1 | - | 4 | 217 | |||
AK165008.1 | - | 1 | 1216 | |||
AK183456.1 | - | 5 | 281 | |||
AK187514.1 | - | 1 | 15 | |||
AK210360.1 | - | 5 | 282 | |||
AK215344.1 | - | 1 | 15 | |||
BC026994.1 | - | 4 | 208 | 5368627 | ||
BC060255.1 | - | 1 | 1089 |