Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026623.3 | NP_080899.1 | 2 | 362 | ||
GenBank mRNA | AK011688.2 | - | 2 | 352 | ||
AK012308.1 | - | 2 | 336 | |||
AK019433.1 | - | 2 | 345 | |||
AK020150.1 | - | 2 | 342 | |||
AK146419.1 | - | 2 | 328 | |||
AK147061.1 | - | 345 | ||||
AK160147.1 | - | 2 | 328 | |||
AK204985.1 | - | 2 | 242 | |||
AK210589.1 | - | 1 | 94 | |||
AK217354.1 | - | 1 | 94 | |||
BC008270.1 | - | 2 | 345 | 2811199 | ||
BC090834.1 | - | 321 | 6855233 |