Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SERPINA1 SIRNA Assays ›
Gene Symbol:
SERPINA1
Gene Name:
serpin family A member 1
Gene Aliases:
A1A, A1AT, AAT, PI, PI1, PRO2275, alpha1AT
Chromosome Location:
Chr.14: 94376747 - 94390692 on Build GRCh38
UniGene ID:
Hs.525557
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
8107
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SERPINA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000295.4
NP_000286.3
2
395
NM_001002235.2
NP_001002235.1
2
374
NM_001002236.2
NP_001002236.1
4
688
NM_001127700.1
NP_001121172.1
2
411
NM_001127701.1
NP_001121173.1
4
707
NM_001127702.1
NP_001121174.1
3
515
NM_001127703.1
NP_001121175.1
4
670
NM_001127704.1
NP_001121176.1
4
667
NM_001127705.1
NP_001121177.1
4
685
NM_001127706.1
NP_001121178.1
3
478
NM_001127707.1
NP_001121179.1
3
475
XM_017021370.1
XP_016876859.1
3
575
GenBank mRNA
AK026174.1
-
2
177
AK315637.1
-
3
431
BC011991.1
-
2
366
3859644
BC015642.2
-
2
161
4644658
BC070163.1
-
2
181
BM844817.1
-
2
393
BM923322.1
-
444
BP296710.1
-
3
418
BP300943.1
-
3
436
BP301274.1
-
3
437
BT019455.1
-
1
134
BX161449.1
-
2
301
BX247968.1
-
4
615
BX248002.1
-
1
143
BX248257.1
-
2
168
BX477352.1
-
2
387
DA427262.1
-
2
354
DA433050.1
-
2
388
DA966896.1
-
2
395
DQ682455.1
-
1
134
GU727620.1
-
1
135
K01396.1
-
1
153
KU178257.1
-
1
134
KU178258.1
-
1
134
M11465.1
-
1
145
X01683.1
-
1
172
Back To Top
More Information
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
platelet degranulation
ER to Golgi vesicle-mediated transport
acute-phase response
blood coagulation
negative regulation of endopeptidase activity
COPII vesicle coating
glycoprotein binding
protease binding
serine-type endopeptidase inhibitor activity
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-tl62x:80/100.66.78.111:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline