Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdipt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026638.3 | NP_080914.1 | 6 | 1572 | ||
GenBank mRNA | AK076974.1 | - | 6 | 1572 | ||
AK079226.1 | - | 6 | 1049 | |||
AK089109.1 | - | 6 | 1138 | |||
AK132286.1 | - | 6 | 1158 | |||
AK193896.1 | - | 1 | 31 | |||
AK197412.1 | - | 1 | 31 | |||
AK198560.1 | - | 1 | 31 | |||
BC021473.1 | - | 6 | 1100 | 5343411 | ||
BC024413.1 | - | 6 | 912 | 4973307 | ||
BC032182.1 | - | 6 | 1109 | 5252470 |