Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177576.1 | NP_001171047.1 | 5 | 466 | ||
NM_026646.3 | NP_080922.1 | 5 | 708 | |||
GenBank mRNA | AK010193.1 | - | 5 | 454 | ||
AK018760.1 | - | 5 | 472 | |||
AK046874.1 | - | 5 | 453 | |||
AK048389.1 | - | 5 | 437 | |||
AK049313.1 | - | 5 | 437 | |||
AK155276.1 | - | 5 | 460 | |||
AK158985.1 | - | 1 | 2635 | |||
AK162400.1 | - | 5 | 419 | |||
BC037949.1 | - | 5 | 465 | 5367108 | ||
BC050887.1 | - | 5 | 661 |