Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026658.2 | NP_080934.2 | 4 | 687 | ||
GenBank mRNA | AF369902.1 | - | 4 | 677 | ||
AK019088.1 | - | 4 | 687 | |||
AK218248.1 | - | 3 | 213 | |||
AK219556.1 | - | 3 | 213 | |||
BC063256.1 | - | 4 | 673 | |||
CF173757.1 | - | 3 | 222 |