Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000301.3 | NP_000292.1 | 2 | 250 | ||
NM_001168338.1 | NP_001161810.1 | 2 | 250 | |||
GenBank mRNA | AK298338.1 | - | 2 | 199 | ||
AK308036.1 | - | 2 | 201 | |||
AK312966.1 | - | 2 | 201 | |||
BC060513.1 | - | 2 | 176 | 4609795 | ||
DR004070.1 | - | 2 | 257 | |||
M74220.1 | - | 2 | 187 | |||
X05199.1 | - | 2 | 192 |
Gene Symbol: PLGLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA634686.1 | - | 2 | 198 |