Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026718.2 | NP_080994.2 | 15 | 2377 | ||
GenBank mRNA | AK003154.1 | - | 1 | 553 | ||
AK039432.1 | - | 15 | 2572 | |||
AK156539.1 | - | 15 | 2570 | |||
AK162340.1 | - | 7 | 1398 | |||
BC003286.1 | - | 5 | 1093 | |||
BC027765.1 | - | 13 | 2041 | |||
BC037744.1 | - | 1 | 309 | |||
BC049187.1 | - | 15 | 2546 | |||
BC057884.1 | - | 4 | 957 | |||
BC061692.1 | - | 15 | 2377 | 6308996 | ||
BC083117.1 | - | 1 | 309 | |||
BC108413.1 | - | 15 | 2840 | 6487251 |