Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000305.2 | NP_000296.2 | 2, 3 | 268 | ||
NM_001018161.1 | NP_001018171.1 | 2, 3 | 268 | |||
XM_005250453.1 | XP_005250510.1 | 2, 3 | 224 | |||
XM_017012357.1 | XP_016867846.1 | 1, 2 | 73 | |||
XM_017012358.1 | XP_016867847.1 | 1, 2 | 73 | |||
GenBank mRNA | AB102891.1 | - | 2, 3 | 147 | ||
AF001601.1 | - | 2, 3 | 179 | |||
AK291103.1 | - | 2, 3 | 190 | |||
AK296029.1 | - | 2, 3 | 183 | |||
AK296636.1 | - | 2, 3 | 224 | |||
AK301313.1 | - | 2, 3 | 395 | |||
AK308065.1 | - | 2, 3 | 198 | |||
AK315209.1 | - | 2, 3 | 216 | |||
BC009728.2 | - | 2, 3 | 224 | |||
BC040010.1 | - | 2, 3 | 171 | 6056277 | ||
BC046160.1 | - | 2, 3 | 187 | 5393700 | ||
CB961097.1 | - | 297 | ||||
L48513.1 | - | 2, 3 | 164 |