Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdipt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026638.3 | NP_080914.1 | 6 | 1846 | ||
GenBank mRNA | AK076974.1 | - | 6 | 1846 | ||
AK079226.1 | - | 6 | 1323 | |||
AK089109.1 | - | 6 | 1412 | |||
AK132286.1 | - | 6 | 1432 | |||
BC021473.1 | - | 6 | 1374 | 5343411 | ||
BC024413.1 | - | 6 | 1186 | 4973307 | ||
BC032182.1 | - | 6 | 1383 | 5252470 |