Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000309.3 | NP_000300.1 | 13 | 1593 | ||
NM_001122764.1 | NP_001116236.1 | 13 | 1654 | |||
XM_006711404.3 | XP_006711467.2 | 13 | 1536 | |||
XM_011509671.1 | XP_011507973.1 | 13 | 1490 | |||
XM_011509672.2 | XP_011507974.1 | 12 | 1599 | |||
XM_011509675.2 | XP_011507977.1 | 12 | 1541 | |||
XM_017001559.1 | XP_016857048.1 | 12 | 1605 | |||
XM_017001560.1 | XP_016857049.1 | 12 | 1521 | |||
XM_017001565.1 | XP_016857054.1 | 13 | 1578 | |||
XM_017001566.1 | XP_016857055.1 | 13 | 1687 | |||
XM_017001567.1 | XP_016857056.1 | 12 | 1480 | |||
XM_017001568.1 | XP_016857057.1 | 13 | 1668 | |||
XM_017001569.1 | XP_016857058.1 | 12 | 1422 | |||
GenBank mRNA | AK302298.1 | - | 12 | 1464 | ||
BC002357.2 | - | 13 | 1574 | 2821983 | ||
D38537.1 | - | 13 | 1641 | |||
U26446.1 | - | 13 | 1555 |