Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam111a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026640.2 | NP_080916.1 | 4 | 655 | ||
GenBank mRNA | AK018830.1 | - | 4 | 664 | ||
AK019499.1 | - | 4 | 656 | |||
AK045189.1 | - | 4 | 657 | |||
AK142496.1 | - | 3 | 455 | |||
AK172402.1 | - | 4 | 668 | |||
BC038020.1 | - | 4 | 649 | 3599082 |