Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mogat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026713.3 | NP_080989.2 | 5 | 629 | ||
GenBank mRNA | AF384162.1 | - | 4 | 506 | ||
AK002693.1 | - | 4 | 578 | |||
BC031920.1 | - | 653 | ||||
BC106135.1 | - | 4 | 577 | 5041577 |