Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026823.2 | NP_081099.1 | 2 | 279 | ||
GenBank mRNA | AB185207.1 | - | 2 | 135 | ||
BC018479.1 | - | 2 | 279 | 3661329 |