Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026917.5 | NP_081193.2 | 3 | 564 | ||
GenBank mRNA | AB088044.1 | - | 2 | 330 | ||
AK007355.1 | - | 4 | 1036 | |||
AK018755.1 | - | 3 | 530 | |||
AK046166.1 | - | 3 | 564 | |||
AK080541.1 | - | 3 | 530 | |||
AK169197.1 | - | 511 | ||||
AK179984.1 | - | 3 | 97 | |||
AY166672.1 | - | 2 | 330 | |||
BC005531.1 | - | 4 | 591 | |||
BC025126.1 | - | 3 | 549 | 3669723 | ||
BC052464.1 | - | 3 | 469 | 6834763 |