Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026921.4 | NP_081197.1 | 3 | 304 | ||
GenBank mRNA | AK007415.1 | - | 3 | 285 | ||
AK045175.1 | - | 3 | 288 | |||
AK090000.1 | - | 3 | 305 | |||
AK153820.1 | - | 3 | 251 | |||
AK169028.1 | - | 3 | 275 | |||
BC018547.1 | - | 3 | 277 | 2615786 | ||
BC085482.1 | - | 3 | 290 | 6830054 |