Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810009N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026939.1 | NP_081215.1 | 1473 | |||
GenBank mRNA | AK007409.1 | - | 1473 | |||
AK084779.1 | - | 959 |
Gene Symbol: 2510022D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009538.1 | - | 760 | |||
AK010990.1 | - | 1080 |