Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex11g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026951.2 | NP_081227.1 | 2 | 204 | ||
GenBank mRNA | AK007582.1 | - | 2 | 205 | ||
AK165461.1 | - | 2 | 226 | |||
BC024096.1 | - | 2 | 235 | |||
BC062135.1 | - | 2 | 193 | 6510277 |