Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026963.5 | NP_081239.1 | 3 | 824 | ||
GenBank mRNA | AK007657.1 | - | 3 | 313 | ||
AK146824.1 | - | 3 | 891 | |||
AK210899.1 | - | 3 | 154 | |||
BC023174.1 | - | 3 | 824 | 4952948 | ||
DQ058008.1 | - | 2 | 149 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK161071.1 | - | 406 | Chr. - - [-] on Build |