Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026810.2 | NP_081086.2 | 2 | 252 | ||
GenBank mRNA | AF250844.1 | - | 2 | 168 | ||
AK038265.1 | - | 2 | 245 | |||
AK145646.1 | - | 2 | 198 | |||
AK163861.1 | - | 2 | 186 | |||
AK168849.1 | - | 2 | 191 | |||
BC021815.1 | - | 2 | 206 | 4020489 | ||
BY327218.1 | - | 2 | 252 |