Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMBS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000190.3 | NP_000181.2 | 9 | 708 | ||
NM_001024382.1 | NP_001019553.1 | 9 | 610 | |||
NM_001258208.1 | NP_001245137.1 | 9 | 708 | |||
NM_001258209.1 | NP_001245138.1 | 9 | 799 | |||
XM_005271531.1 | XP_005271588.1 | 9 | 786 | |||
XM_005271532.1 | XP_005271589.1 | 9 | 764 | |||
XM_005271533.3 | XP_005271590.1 | 8 | 660 | |||
XM_011542796.1 | XP_011541098.1 | 8 | 1040 | |||
XM_017017629.1 | XP_016873118.1 | 9 | 1174 | |||
GenBank mRNA | AK000628.1 | - | 9 | 791 | ||
AK131072.1 | - | 9 | 671 | |||
AK290275.1 | - | 9 | 704 | |||
AK309544.1 | - | 9 | 845 | |||
AK310343.1 | - | 703 | ||||
BC000520.2 | - | 9 | 702 | 2822949 | ||
BC008149.1 | - | 9 | 663 | 4178944 | ||
BC019323.1 | - | 9 | 702 | 2822949 | ||
BX647328.1 | - | 8 | 839 | |||
M60888.1 | - | 1 | 53 | |||
X04217.1 | - | 9 | 581 | |||
X04808.1 | - | 9 | 578 |