Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026850.4 | NP_081126.2 | 3 | 212 | ||
GenBank mRNA | AF110512.1 | - | 2 | 182 | ||
AK078372.1 | - | 3 | 213 | |||
AK168509.1 | - | 3 | 253 | |||
BC005601.1 | - | 3 | 233 | 3483084 |