Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026921.4 | NP_081197.1 | 4 | 866 | ||
GenBank mRNA | AK041946.1 | - | 1 | 65 | ||
AK045175.1 | - | 4 | 850 | |||
AK090000.1 | - | 4 | 867 | |||
AK153820.1 | - | 4 | 813 | |||
AK169028.1 | - | 4 | 837 | |||
BC018547.1 | - | 4 | 839 | 2615786 | ||
BC059872.1 | - | 1 | 2230 | |||
BC085482.1 | - | 4 | 852 | 6830054 |