Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebna1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026932.4 | NP_081208.1 | 2, 3 | 276 | ||
GenBank mRNA | AK007491.1 | - | 2, 3 | 246 | ||
AK134732.1 | - | 2, 3 | 231 | |||
AK167070.1 | - | 2, 3 | 231 | |||
AK168269.1 | - | 2, 3 | 203 | |||
BC054723.1 | - | 2, 3 | 259 | 5695176 | ||
BC062876.1 | - | 2, 3 | 208 | 5686021 | ||
BY324791.1 | - | 2, 3 | 276 |