Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026934.3 | NP_081210.2 | 7 | 910 | ||
GenBank mRNA | AK007452.1 | - | 2 | 141 | ||
AK139511.1 | - | 7 | 1006 | |||
AK152374.1 | - | 7 | 1001 | |||
AK159596.1 | - | 7 | 982 | |||
AK167696.1 | - | 7 | 1003 | |||
BC028787.1 | - | 2 | 349 | 1246186 | ||
BC031845.1 | - | 7 | 910 | 3983096 | ||
BC058229.1 | - | 7 | 837 | 6313954 |