Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026949.3 | NP_081225.1 | 4 | 488 | ||
GenBank mRNA | AK007581.1 | - | 4 | 552 | ||
AK075727.1 | - | 4 | 458 | |||
AK150757.1 | - | 4 | 569 | |||
AK153907.1 | - | 4 | 491 | |||
AK216161.1 | - | 2 | 100 | |||
AY515608.1 | - | 3 | 455 | |||
BC004040.1 | - | 4 | 488 | 3496188 |