Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289535.1 | NP_001276464.1 | 2 | 326 | ||
NM_001289536.1 | NP_001276465.1 | 2 | 326 | |||
NM_026959.3 | NP_081235.2 | 2 | 326 | |||
GenBank mRNA | AK007700.1 | - | 2 | 324 | ||
AK016910.1 | - | 2 | 268 | |||
AK169273.1 | - | 2 | 270 | |||
AK170165.1 | - | 2 | 322 | |||
BC021362.1 | - | 2 | 316 | 5035350 | ||
BY037320.1 | - | 2 | 327 |