Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026960.4 | NP_081236.1 | 3 | 431 | ||
GenBank mRNA | AK007710.1 | - | 3 | 432 | ||
AK088747.1 | - | 3 | 443 | |||
AK165858.1 | - | 3 | 432 | |||
AK171294.1 | - | 3 | 433 | |||
BC029813.1 | - | 3 | 384 | 4194837 |