Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026992.3 | NP_081268.1 | 3 | 692 | ||
GenBank mRNA | AK006521.1 | - | 3 | 693 | ||
AY028460.1 | - | 3 | 374 | |||
BC030072.1 | - | 3 | 400 | 3376358 | ||
BC091774.1 | - | 3 | 258 | 5150845 |