Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026810.2 | NP_081086.2 | 6 | 580 | ||
GenBank mRNA | AF250844.1 | - | 6 | 496 | ||
AK038265.1 | - | 6 | 573 | |||
AK145646.1 | - | 6 | 527 | |||
AK168849.1 | - | 6 | 519 | |||
BC021815.1 | - | 6 | 534 | 4020489 |